Hexaphenylcyclotrisiloxane ≥96.0%
Supplier: TCI America
H1248-5G
TCH1248-5GEA
176
USD
TCH1248-5G
Hexaphenylcyclotrisiloxane ≥96.0%
Hexaphenylcyclotrisiloxane
CAS Number: 512-63-0
MDL Number: MFCD00023110
Molecular Formula: C36H30O3Si3
Molecular Weight: 594.89
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 300
Melting point (°C): 190
Formula:
C₃₆H₃₀O₃Si₃ Boiling Pt: 300 °C Melting Pt: 190 °C |
MDL Number:
MFCD00023110 CAS Number: 512-63-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...