Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, Order Status will not be shown from 8:00am – 10:00am CEST Sunday.

4-Hydroxy-3-nitropyridine ≥98.0% (by GC, titration analysis)

Supplier: TCI America


H1171-5G H1171-25G
TCH1171-5GEA 57.23 USD InStock
TCH1171-5G TCH1171-25G
4-Hydroxy-3-nitropyridine ≥98.0% (by GC, titration analysis)
4-Hydroxy-3-nitropyridine
CAS Number: 5435-54-1
Molecular Formula: C5H4N2O3
Molecular Weight: 140.10
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 284

Other Product Names / Synonyms: 3-Nitro-4-pyridinol


Formula: C₅H₄N₂O₃
MW: 140.1 g/mol
Melting Pt: 284 °C
MDL Number: MFCD00128874
CAS Number: 5435-54-1
EINECS: 000-000-0
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty