Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Heptadecafluoro-10-iododecane ≥98.0%

Supplier: TCI America

Synonyms: 1-Iodo-1H,1H,2H,2H-perfluorodecane1H,1H,2H,2H-Perfluorodecyliodide1H,1H,2H,2H-Perfluoro-1-iododecane

H1084-5G H1084-25G
TCH1084-5GEA 24.97 USD InStock
TCH1084-5G TCH1084-25G
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Heptadecafluoro-10-iododecane ≥98.0%
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8-Heptadecafluoro-10-iododecane
CAS Number: 2043-53-0
MDL Number: MFCD00039411
Molecular Formula: C10H4F17I
Molecular Weight: 574.02
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 96
Melting point (°C): 58

Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks, Materials Chemistry, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: 1H,1H,2H,2H-Perfluorodecyl Iodide, 1H,1H,2H,2H-Heptadecafluoro-1-iododecane, 1H,1H,2H,2H-Perfluoro-1-iododecane


Formula: C₁₀H₄F₁₇I
MW: 574,02 g/mol
Boiling Pt: 96 °C
Melting Pt: 58 °C
Density: 1.88 g/cm³ (60 °C)
Storage Temperature: ambient
MDL Number: MFCD00039411
CAS Number: 2043-53-0
EINECS: 218-053-5
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty