4'-Hydroxyflavanone ≥98.0% (by HPLC)
Supplier: TCI America
H1026-1G
TCH1026-1GEA
118
USD
TCH1026-1G
4'-Hydroxyflavanone ≥98.0% (by HPLC)
4'-Hydroxyflavanone
CAS Number: 6515-37-3
MDL Number: MFCD00017705
Molecular Formula: C15H12O3
Molecular Weight: 240.26
Purity/Analysis Method:
Form: Crystal
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...