Isoscopoletin (6-hydroxy-4-methylcoumarin) ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
6-Hydroxy-4-methylcoumarin
, 6-Hydroxy-4-methyl-2H-chromen-2-one
H1005-25G
H1005-5G
TCH1005-25GEA
126.48
USD
TCH1005-25G
TCH1005-5G
Isoscopoletin (6-hydroxy-4-methylcoumarin) ≥98.0% (by GC, titration analysis)
Isoscopoletin (6-Hydroxy-4-methylcoumarin)
CAS Number: 2373-31-1
MDL Number: MFCD00016967
Molecular Formula: C10H8O3
Molecular Weight: 176.17
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 248
Formula:
C₁₀H₈O₃ MW: 176.17 g/mol Melting Pt: 248 °C |
MDL Number:
MFCD00016967 CAS Number: 2373-31-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...