(Hydrazinocarbonyl)ferrocene ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Ferrocenecarbohydrazide

H0941-1G
TCH0941-001GEA 479 USD
TCH0941-001G
(Hydrazinocarbonyl)ferrocene ≥95.0% (by HPLC, titration analysis)
(Hydrazinocarbonyl)ferrocene

[for HPLC Labeling]
CAS Number: 12153-28-5
MDL Number: MFCD01321175
Molecular Formula: C11H12FeN2O
Molecular Weight: 244.08
Purity/Analysis Method: 95.0% (HPLC,T)
Form: Crystal


Product Categories: Analytical Science, Electrochemical Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: Ferrocenecarbohydrazide

Formula: C₁₁H₁₂FeN₂O
MW: 244.08 g/mol
MDL Number: MFCD01321175
CAS Number: 12153-28-5
https://us.vwr.com/stibo/web/std.lang.all/09/61/28970961.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR