(S)-3-Hydroxy-γ-butyrolactone ≥95.0%
Synonyms:
(S)-4-Hydroxytetrahydrofuran-2-one
H0939-5G
H0939-1G
TCH0939-005GEA
167.28
USD
TCH0939-005G
TCH0939-1G
(S)-3-Hydroxy-γ-butyrolactone ≥95.0%
(S)-3-Hydroxy-gamma-butyrolactone
CAS Number: 7331-52-4
MDL Number: MFCD00211247
Molecular Formula: C4H6O3
Molecular Weight: 102.09
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 100
Flash Point (°C): 109
Specific Gravity (20/20): 1.31
Specific rotation [a]20/D: -83 deg (C=2, EtOH)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-4-Hydroxytetrahydrofuran-2-one
Formula:
C₄H₆O₃ MW: 102.09 g/mol Boiling Pt: 100 °C Density: 1.31 (20 °C) Flash Pt: 109 °C |
MDL Number:
MFCD00211247 CAS Number: 7331-52-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...