4-Methoxysalicylaldehyde ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Hydroxy-4-methoxybenzaldehyde

H0699-25G H0699-5G
TCH0699-5GEA 65 USD
TCH0699-5G TCH0699-25G
4-Methoxysalicylaldehyde ≥98.0% (by GC, titration analysis)
4-Methoxysalicylaldehyde

CAS Number: 673-22-3
MDL Number: MFCD00003327
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 124
Melting point (°C): 42
Flash Point (°C): 110


Other Product Names / Synonyms: 2-Hydroxy-p-anisaldehyde, 4-Methoxysalicylaldehyde

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 124 °C
Melting Pt: 42 °C
Flash Pt: 110 °C
MDL Number: MFCD00003327
CAS Number: 673-22-3
https://us.vwr.com/stibo/web/std.lang.all/07/43/28970743.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR