DL-Homocysteinethiolactone hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
3-Aminodihydrothiophen-2(3H)-one hydrochloride
, DL-3-Aminotetrahydrothiophen-2-one Hydrochloride
H0646-25G
TCH0646-025GEA
29.44
USD
TCH0646-025G
DL-Homocysteinethiolactone hydrochloride ≥98.0% (by titrimetric analysis)
DL-Homocysteinethiolactone hydrochloride
CAS Number: 6038-19-3
MDL Number: MFCD00012724
Molecular Formula: C4H7NOS
Molecular Weight: 153.62
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: DL-3-Aminotetrahydrothiophen-2-one Hydrochloride
Formula:
C₄H₇NOS MW: 153.62 g/mol |
MDL Number:
MFCD00012724 CAS Number: 6038-19-3 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...