Hexahydro-1,3,5-triphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)

Synonyms: 1,3,5-Triphenyl-1,3,5-triazinane

H0639-25G
TCH0639-025GEA 37.08 USD
TCH0639-025G
Hexahydro-1,3,5-triphenyl-1,3,5-triazine ≥98.0% (by HPLC, total nitrogen)
Hexahydro-1,3,5-triphenyl-1,3,5-triazine

CAS Number: 91-78-1
MDL Number: MFCD00043590
Molecular Formula: C21H21N3
Molecular Weight: 315.42
Purity/Analysis Method: 98.0% (HPLC,N)
Form: Crystal
Flash Point (°C): 100
Storage Temperature: 0-10°C

Formula: C₂₁H₂₁N₃
MW: 315.42 g/mol
Flash Pt: 100 °C
MDL Number: MFCD00043590
CAS Number: 91-78-1
https://us.vwr.com/stibo/web/std.lang.all/06/51/28970651.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR