4-Hydroxyphthalic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
H0609-5G
TCH0609-5GEA
109
USD
TCH0609-5G
4-Hydroxyphthalic acid ≥98.0% (by titrimetric analysis)
4-Hydroxyphthalic acid
CAS Number: 610-35-5
MDL Number: MFCD00013984
Molecular Formula: C8H6O5
Molecular Weight: 182.13
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 198
Formula:
C₈H₆O₅ MW: 182.13 g/mol Melting Pt: 198 °C |
MDL Number:
MFCD00013984 CAS Number: 610-35-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...