Hexyltrimethylammonium bromide ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
(1-Hexyl)trimethylammonium bromide
, Hexyltrimethylammonium Bromide
H0534-500G
H0534-25G
TCH0534-025GEA
110.29
USD
TCH0534-025G
TCH0534-500G
Hexyltrimethylammonium bromide ≥98.0% (by HPLC, titration analysis)
Hexyltrimethylammonium bromide
CAS Number: 2650-53-5
MDL Number: MFCD00051872
Molecular Formula: C9H22BrN
Molecular Weight: 224.19
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 185
Formula:
C₉H₂₂BrN MW: 224.18 g/mol Melting Pt: 185 °C |
MDL Number:
MFCD00051872 CAS Number: 2650-53-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...