Hydroxylamine-O-sulfonic acid ≥90.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Amino hydrogen sulphate

H0530-100G H0530-500G H0530-25G
TCH0530-25GEA 120.03 USD
TCH0530-25G TCH0530-500G TCH0530-100G
Hydroxylamine-O-sulfonic acid ≥90.0% (by titrimetric analysis)
Hydroxylamine-O-sulphonic acid

CAS Number: 2950-43-8
MDL Number: MFCD00011604
Molecular Formula: H3NO4S
Molecular Weight: 113.09
Purity/Analysis Method: 90.0% (T)
Form: Crystal

Formula: H₃NO₄S
MW: 113.09
MDL Number: MFCD00011604
CAS Number: 2950-43-8
UN: 3260
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/05/39/28970539.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR