Ferrocenemethanol ≥95.0%
Supplier: TCI America
Synonyms:
Hydroxymethylferrocene
H0464-1G
H0464-5G
TCH0464-001GEA
102.72
USD
TCH0464-001G
TCH0464-5G
Ferrocenemethanol ≥95.0%
Ferrocenemethanol
CAS Number: 1273-86-5
MDL Number: MFCD00040878
Molecular Formula: C11H12FeO
Molecular Weight: 216.06
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 78
Other Product Names / Synonyms: Ferrocenylmethanol
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...