Hexamethylene oxide ≥98.0%

Supplier: TCI America

Synonyms: OxepaneOxacycloheptane

H0446-25ML
TCH0446-025MLEA 430 USD
TCH0446-025ML
Hexamethylene oxide ≥98.0%
Hexamethylene oxide

CAS Number: 592-90-5
MDL Number: MFCD00003266
Molecular Formula: C6H12O
Molecular Weight: 100.16
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 119
Flash Point (°C): 10
Specific Gravity (20/20): 0.90


Other Product Names / Synonyms: Oxacycloheptane, Oxepane

Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 119 °C
Density: 0.90 (20 °C)
Flash Pt: 10 °C
MDL Number: MFCD00003266
CAS Number: 592-90-5
https://us.vwr.com/stibo/web/std.lang.all/04/62/28970462.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR