Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, Order Status will not be shown from 8:00am – 10:00am CEST Sunday.

(1H-Imidazol-4-yl)methanol hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-(Hydroxymethyl)imidazole hydrochloride4(5)-Imidazolemethanol hydrochloride

H0414-1G
TCH0414-001GEA 173.75 USD InStock
TCH0414-001G
(1H-Imidazol-4-yl)methanol hydrochloride ≥98.0% (by titrimetric analysis)
(1H-Imidazol-4-yl)methanol hydrochloride
CAS Number: 32673-41-9
MDL Number: MFCD00012697
Molecular Formula: C4H6N2O
Molecular Weight: 134.56
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 108
Storage Temperature: 0-10°C


Formula: C₄H₇ClN₂O
MW: 134,57
Melting Pt: 108 °C
MDL Number: MFCD00012697
CAS Number: 32673-41-9
EINECS: 251-150-0
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty