N-(4-Hydroxyphenyl)glycine ≥97.0% (by HPLC, titration analysis)

Supplier: TCI America

H0292-25G
TCH0292-025GEA 124 USD
TCH0292-025G
N-(4-Hydroxyphenyl)glycine ≥97.0% (by HPLC, titration analysis)
N-(4-Hydroxyphenyl)glycine

CAS Number: 122-87-2
MDL Number: MFCD00002689
Molecular Formula: C8H9NO3
Molecular Weight: 167.16
Purity/Analysis Method: 97.0% (HPLC,T)
Form: Crystal

Formula: C₈H₉NO₃
MDL Number: MFCD00002689
CAS Number: 122-87-2
https://us.vwr.com/stibo/web/std.lang.all/03/37/28970337.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR