Formaldehyde sodium bisulfite ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Sodium HydroxymethanesulphonateHydroxymethanesulphonic acid sodium saltSodium HydroxymethanesulfonateHydroxymethanesulfonic acid sodium salt

H0260-25G
TCH0260-025GEA 43.98 USD
TCH0260-025G
Formaldehyde sodium bisulfite ≥97.0% (by titrimetric analysis)
Formaldehyde sodium bisulfite

CAS Number: 870-72-4
MDL Number: MFCD00044664
Molecular Formula: CH4O4S
Molecular Weight: 134.08
Purity/Analysis Method: 97.0% (T)
Form: Crystal


Other Product Names / Synonyms: Sodium Hydroxymethanesulfonate, Hydroxymethanesulfonic Acid Sodium Salt

Formula: CH₄O₄S
MW: 134.08 g/mol
MDL Number: MFCD00044664
CAS Number: 870-72-4
https://us.vwr.com/stibo/web/std.lang.all/02/98/28970298.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR