Ethyl-2-hydroxyisobutyrate ≥98.0%

Supplier: TCI America

H0255-25G H0255-250G
TCH0255-250GEA 275.07 USD
TCH0255-250G TCH0255-025G
Ethyl-2-hydroxyisobutyrate ≥98.0%
Ethyl 2-hydroxyisobutyrate

CAS Number: 80-55-7
MDL Number: MFCD00004458
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 41
Specific Gravity (20/20): 0.99


Other Product Names / Synonyms: 2-Hydroxyisobutyric Acid Ethyl Ester

Formula: C₆H₁₂O₃
MW: 132.16
Boiling Pt: 150 °C
Density: 0.99 (20 °C)
Flash Pt: 41 °C
MDL Number: MFCD00004458
CAS Number: 80-55-7
UN: 3272
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/02/95/28970295.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR