TCIAH0146

H0146-1G
TCH0146-001GEA 19.7 USD
TCH0146-001G
TCIAH0146
CAS Number: 56-92-8
MDL Number: MFCD00012703
Molecular Formula: C5H9N3
Molecular Weight: 184.06
Purity/Analysis Method: 99.0% (T)
Form: Crystal
Melting point (°C): 246
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR