Hexanophenone ≥98.0%

Supplier: TCI America

H0116-25G H0116-500G
TCH0116-025GEA 71 USD
TCH0116-025G TCH0116-500G
Hexanophenone ≥98.0%
Hexanophenone

CAS Number: 942-92-7
MDL Number: MFCD00009512
Molecular Formula: C12H16O
Molecular Weight: 176.26
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 25


Other Product Names / Synonyms: Amyl Phenyl Ketone, Caprophenone

Formula: C₁₂H₁₆O
MW: 176.26 g/mol
Flash Pt: 110 °C
MDL Number: MFCD00009512
CAS Number: 942-92-7
https://us.vwr.com/stibo/web/std.lang.all/01/81/28970181.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR