Harmine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Banisterine

H0001-1G
TCH0001-001GEA 145 USD
TCH0001-001G
Harmine ≥98.0% (by titrimetric analysis)
Harmine

CAS Number: 442-51-3
MDL Number: MFCD00004958
Molecular Formula: C13H12N2O
Molecular Weight: 212.25
Purity/Analysis Method: 98.0% (T)
Form: Crystal


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Indole Alkaloids
Other Product Names / Synonyms: Banisterine

Formula: C₁₃H₁₂N₂O
MDL Number: MFCD00004958
CAS Number: 442-51-3
https://us.vwr.com/stibo/web/std.lang.all/00/82/28970082.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR