Calcium-DL-glycerate hydrate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
DL-Glyceric Acid Calcium Salt
, Calcium 2,3-Dihydroxypropionate
, 2,3-Dihydroxypropionic Acid Calcium Salt
G0232-10G
TCG0232-010GEA
355.25
USD
TCG0232-010G
Calcium-DL-glycerate hydrate ≥98.0% (by titrimetric analysis)
Calcium-DL-glycerate hydrate
CAS Number: 67525-74-0
MDL Number: MFCD00149289
Molecular Formula: C3H6O4
Molecular Weight: 250.22
Purity/Analysis Method:
Form: Crystal
Other Product Names / Synonyms: Calcium 2,3-Dihydroxypropionate, DL-Glyceric Acid Calcium Salt, 2,3-Dihydroxypropionic Acid Calcium Salt
Formula:
C₃H₆O₄ MW: 250.22 g/mol |
MDL Number:
MFCD00149289 CAS Number: 67525-74-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...