2-Fluoro-4-iodotoluene ≥95.0%

Supplier: TCI America

Synonyms: 1-Fluoro-5-iodo-2-methylbenzene2-Fluoro-4-iodo-1-methylbenzene

F0720-5G F0720-25G
TCF0720-5GEA 43 USD
TCF0720-5G TCF0720-25G
2-Fluoro-4-iodotoluene ≥95.0%
2-Fluoro-4-iodotoluene

CAS Number: 39998-81-7
MDL Number: MFCD00001045
Molecular Formula: C7H6FI
Molecular Weight: 236.03
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 208
Flash Point (°C): 79
Specific Gravity (20/20): 1.79

Formula: C₇H₆FI
MW: 236.03 g/mol
Boiling Pt: 208 °C
Density: 1.79 (20 °C)
Flash Pt: 79 °C
MDL Number: MFCD00001045
CAS Number: 39998-81-7
https://us.vwr.com/stibo/web/std.lang.all/92/53/28969253.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR