Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluorophenyl isothiocyanate ≥97.0%

Supplier: TCI America


F0711-5G F0711-25G
TCF0711-5GEA 87.4 USD InStock
TCF0711-5G TCF0711-25G
2-Fluorophenyl isothiocyanate ≥97.0%
2-Fluorophenyl isothiocyanate
CAS Number: 38985-64-7
MDL Number: MFCD00004800
Molecular Formula: C7H4FNS
Molecular Weight: 153.17
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 104
Flash Point (°C): 97
Specific Gravity (20/20): 1.25

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Isothiocyanic Acid 2-Fluorophenyl Ester


Formula: C₇H₄FNS
MW: 153,18 g/mol
Boiling Pt: 104 °C
Melting Pt: 24 to 26 °C
Density: 1.25 (20 °C)
Flash Pt: 97 °C
MDL Number: MFCD00004800
CAS Number: 38985-64-7
EINECS: 254-237-1
UN: 1760
ADR: 8,II
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty