1-Furfurylpyrrole ≥98.0%

Supplier: TCI America

Synonyms: 1-(Furan-2-ylmethyl)-1H-pyrrole

F0700-5G
TCF0700-5GEA 168.46 USD
TCF0700-5G
1-Furfurylpyrrole ≥98.0%
1-Furfurylpyrrole

CAS Number: 1438-94-4
MDL Number: MFCD00010080
Molecular Formula: C9H9NO
Molecular Weight: 147.18
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 79
Flash Point (°C): 93
Specific Gravity (20/20): 1.08
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-(1-Pyrrolylmethyl)furan

Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 79 °C
Density: 1.08 (20 °C)
Flash Pt: 93 °C
MDL Number: MFCD00010080
CAS Number: 1438-94-4
https://us.vwr.com/stibo/web/std.lang.all/92/38/28969238.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR