Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoro-6-nitrotoluene ≥98.0%

Supplier: TCI America


F0696-5G F0696-25G
TCF0696-5GEA 30.17 USD InStock
TCF0696-5G TCF0696-25G
2-Fluoro-6-nitrotoluene ≥98.0%
2-Fluoro-6-nitrotoluene
CAS Number: 769-10-8
MDL Number: MFCD00007197
Molecular Formula: C7H6FNO2
Molecular Weight: 155.13
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 97
Melting point (°C): 7
Flash Point (°C): 89
Specific Gravity (20/20): 1.28


Formula: C₇H₆FNO₂
MW: 155,13 g/mol
Boiling Pt: 97 °C
Melting Pt: 7 °C
Density: 1.28 (20 °C)
Flash Pt: 89 °C
MDL Number: MFCD00007197
CAS Number: 769-10-8
EINECS: 212-203-3
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty