Fluoromethyl-1,1,1,3,3,3-hexafluoroisopropyl ether ≥98.0%

Supplier: TCI America

F0691-5G
TCF0691-5GEA 205.03 USD
TCF0691-5G
Fluoromethyl-1,1,1,3,3,3-hexafluoroisopropyl ether ≥98.0%
Fluoromethyl-1,1,1,3,3,3-hexafluoroisopropyl ether

CAS Number: 28523-86-6
MDL Number: MFCD00153189
Molecular Formula: C4H3F7O
Molecular Weight: 200.06
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 58
Specific Gravity (20/20): 1.52
Storage Temperature: 0-10°C

Formula: C₄H₃F₇O
MW: 200.06 g/mol
Boiling Pt: 58 °C
Density: 1.52 (20 °C)
MDL Number: MFCD00153189
CAS Number: 28523-86-6
https://us.vwr.com/stibo/web/std.lang.all/92/29/28969229.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR