4-Fluoro-3-methoxybenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Methoxy-4-fluorobenzoic acid

F0681-5G F0681-1G
TCF0681-5GEA 204 USD
TCF0681-5G TCF0681-1G
4-Fluoro-3-methoxybenzoic acid ≥98.0% (by GC, titration analysis)
4-Fluoro-3-methoxybenzoic acid

CAS Number: 82846-18-2
MDL Number: MFCD00272134
Molecular Formula: C8H7FO3
Molecular Weight: 170.14
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 207


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluoro-m-anisic Acid

Formula: C₈H₇FO₃
MW: 170.14 g/mol
Melting Pt: 207 °C
MDL Number: MFCD00272134
CAS Number: 82846-18-2
https://us.vwr.com/stibo/web/std.lang.all/92/25/28969225.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR