Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4-Dimethoxy-2-fluorobenzene ≥97.0%

Supplier: TCI America


F0633-5G
TCF0633-5GEA 89.48 USD InStock
TCF0633-5G
1,4-Dimethoxy-2-fluorobenzene ≥97.0%
1,4-Dimethoxy-2-fluorobenzene
CAS Number: 82830-49-7
MDL Number: MFCD00042487
Molecular Formula: C8H9FO2
Molecular Weight: 156.16
Purity/Analysis Method: 97.0% (GC)
Boiling point (°C): 121
Freezing point (°C): 24


Formula: C₈H₉FO₂
MW: 156,16 g/mol
Boiling Pt: 121 °C
Melting Pt: 23 to 26 °C
MDL Number: MFCD00042487
CAS Number: 82830-49-7
EINECS: 000-000-0
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty