Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(4-Fluorophenyl)ethanol ≥97.0%

Supplier: TCI America

Synonyms: 4-Fluorophenethyl alcohol

F0624-5G
TCF0624-5GEA 58.27 USD InStock
TCF0624-5G
2-(4-Fluorophenyl)ethanol ≥97.0%
2-(4-Fluorophenyl)ethanol
CAS Number: 7589-27-7
MDL Number: MFCD00002898
Molecular Formula: C8H9FO
Molecular Weight: 140.16
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 118
Flash Point (°C): 86
Specific Gravity (20/20): 1.13


Formula: C₈H₉FO
MW: 140,16 g/mol
Boiling Pt: 118 °C
Density: 1,13 (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00002898
CAS Number: 7589-27-7
EINECS: 231-499-5
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty