2-Fluorobenzhydrazide ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-Fluorobenzoic hydrazide
, 2-Fluorobenzoic acid hydrazide
, 2-Fluorobenzene-1-carbohydrazide
, 2-Fluorobenzohydrazide
CAS Number: 446-24-2
MDL Number: MFCD00025112
Molecular Formula: C7H7FN2O
Molecular Weight: 154.14
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 73
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluorobenzoylhydrazine, 2-Fluorobenzoic Hydrazide
Formula:
C₇H₇FN₂O MW: 154.14 g/mol Melting Pt: 73 °C |
MDL Number:
MFCD00025112 CAS Number: 446-24-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...