Fmoc-D-Pro-OH ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: N-Fmoc-D-proline

F0606-5G
TCF0606-5GEA 123 USD
TCF0606-5G
Fmoc-D-Pro-OH ≥98.0% (by HPLC)
Fmoc-D-Pro-OH

CAS Number: 101555-62-8
MDL Number: MFCD00077067
Molecular Formula: C20H19NO4
Molecular Weight: 337.38
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: -32 deg (C=1, DMF)


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-D-proline, Fmoc-D-Pro-OH

Formula: C₂₀H₁₉NO₄
MDL Number: MFCD00077067
CAS Number: 101555-62-8
https://us.vwr.com/stibo/web/std.lang.all/91/58/28969158.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR