Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

S-Furfuryl thiopropionate ≥98.0%

Supplier: TCI America


F0578-25G
TCF0578-25GEA 62.43 USD InStock
TCF0578-25G
S-Furfuryl thiopropionate ≥98.0%
S-Furfuryl thiopropionate
CAS Number: 59020-85-8
MDL Number: MFCD00040264
Molecular Formula: C8H10O2S
Molecular Weight: 170.23
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 98
Specific Gravity (20/20): 1.12

Other Product Names / Synonyms: Thiopropionic Acid S-Furfuryl Ester


Formula: C₈H₁₀O₂S
MW: 170,23
Density: 1,12 (20 °C)
Flash Pt: 98 °C
MDL Number: MFCD00040264
CAS Number: 59020-85-8
EINECS: 261-562-2
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty