Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoronicotinic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Fluoro-3-pyridinecarboxylic acid2-Fluoropyridine-3-carboxylic acid

F0575-5G F0575-1G
TCF0575-5GEA 158.15 USD InStock
TCF0575-5G TCF0575-1G
2-Fluoronicotinic acid ≥98.0% (by GC, titration analysis)
2-Fluoronicotinic acid
CAS Number: 393-55-5
MDL Number: MFCD00040744
Molecular Formula: C6H4FNO2
Molecular Weight: 141.10
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 165

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluoro-3-pyridinecarboxylic Acid


Formula: C₆H₄FNO₂
MW: 141,1 g/mol
Melting Pt: 165 °C
MDL Number: MFCD00040744
CAS Number: 393-55-5
EINECS: 206-888-8
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty