4-Fluoro-m-toluic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0560-5G
TCF0560-5GEA 88.31 USD
TCF0560-5G
4-Fluoro-m-toluic acid ≥98.0% (by GC, titration analysis)
4-Fluoro-m-toluic acid

CAS Number: 403-15-6
MDL Number: MFCD01631424
Molecular Formula: C8H7FO2
Molecular Weight: 154.14
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 168


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Fluoro-m-toluic Acid

Formula: C₈H₇FO₂
MW: 154.14 g/mol
Melting Pt: 168 °C
MDL Number: MFCD01631424
CAS Number: 403-15-6
https://us.vwr.com/stibo/web/std.lang.all/91/23/28969123.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR