2'-Fluoroacetanilide ≥98.0%

Supplier: TCI America

F0524-25G
TCF0524-25GEA 53.8 USD
TCF0524-25G
2'-Fluoroacetanilide ≥98.0%
2'-Fluoroacetanilide

CAS Number: 399-31-5
Molecular Formula: C8H8FNO
Molecular Weight: 153.16
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 142
Melting point (°C): 78


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Acetamido-2-fluorobenzene, N-(2-Fluorophenyl)acetamide

Formula: C₈H₈FNO
MW: 153.16 g/mol
Boiling Pt: 142 °C
Melting Pt: 78 °C
MDL Number: MFCD00000315
CAS Number: 399-31-5
https://us.vwr.com/stibo/web/std.lang.all/90/88/28969088.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR