4'-Fluoroacetanilide ≥98.0%

Supplier: TCI America

Synonyms: 4-FluoroacetanilideN-(4-Fluorophenyl)acetamide

F0521-25G
TCF0521-25GEA 148 USD
TCF0521-25G
4'-Fluoroacetanilide ≥98.0%
4'-Fluoroacetanilide

CAS Number: 351-83-7
Molecular Formula: C8H8FNO
Molecular Weight: 153.16
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 154


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-Acetamido-4-fluorobenzene, N-(4-Fluorophenyl)acetamide

Formula: C₈H₈FNO
MW: 153.16 g/mol
Melting Pt: 154 °C
MDL Number: MFCD00016335
CAS Number: 351-83-7
https://us.vwr.com/stibo/web/std.lang.all/90/85/28969085.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR