1-(2-Chloroethyl)-4-fluorobenzene ≥97.0%

Supplier: TCI America

F0478-1G
TCF0478-1GEA 30 USD
TCF0478-1G
1-(2-Chloroethyl)-4-fluorobenzene ≥97.0%
1-(2-Chloroethyl)-4-fluorobenzene

CAS Number: 332-43-4
MDL Number: MFCD00155000
Molecular Formula: C8H8ClF
Molecular Weight: 158.60
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.17


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 1-(2-Chloroethyl)-4-fluorobenzene, 1-Chloro-2-(4-fluorophenyl)ethane, 4-Fluorophenethyl Chloride

Formula: C₈H₈ClF
MW: 158.6 g/mol
Density: 1.17 (20 °C)
MDL Number: MFCD00155000
CAS Number: 332-43-4
https://us.vwr.com/stibo/web/std.lang.all/90/61/28969061.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR