Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(4-Fluorophenyl)indole ≥98.0%

Supplier: TCI America


F0427-5G F0427-25G
76220-068EA 66.95 USD InStock
76220-068 TCF0427-25G
2-(4-Fluorophenyl)indole ≥98.0%
2-(4-Fluorophenyl)indole
CAS Number: 782-17-2
MDL Number: MFCD00068178
Molecular Formula: C14H10FN
Molecular Weight: 211.24
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 190


Formula: C₁₄H₁₀FN
MW: 211,24 g/mol
Melting Pt: 190 °C
MDL Number: MFCD00068178
CAS Number: 782-17-2
EINECS: 000-000-0
UN: 0000
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty