N-Formylglycine ethyl ester ≥98.0%
Supplier: TCI America
Synonyms:
Ethyl Formamidoacetate
F0421-5G
F0421-25G
TCF0421-5GEA
46.67
USD
TCF0421-5G
TCF0421-025G
N-Formylglycine ethyl ester ≥98.0%
N-Formylglycine ethyl ester
CAS Number: 3154-51-6
MDL Number: MFCD00014125
Molecular Formula: C5H9NO3
Molecular Weight: 131.13
Purity/Analysis Method:
Form: Clear Liquid
Specific Gravity (20/20): 1.15
Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: Ethyl Formamidoacetate, Formamidoacetic Acid Ethyl Ester, For-Gly-OEt
Formula:
C₅H₉NO₃ MW: 131.13 g/mol Density: 1.15 (20 °C) |
MDL Number:
MFCD00014125 CAS Number: 3154-51-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...