Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fluorene-2-carbaldehyde ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Fluorene-2-carboxaldehyde

F0333-5G
TCF0333-005GEA 71.79 USD InStock
TCF0333-005G
Fluorene-2-carbaldehyde ≥95.0% (by titrimetric analysis)
Fluorene-2-carbaldehyde
CAS Number: 30084-90-3
MDL Number: MFCD00001123
Molecular Formula: C14H10O
Molecular Weight: 194.23
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Melting point (°C): 84


Formula: C₁₄H₁₀O
MW: 194,23
Melting Pt: 84 °C
MDL Number: MFCD00001123
CAS Number: 30084-90-3
EINECS: 250-035-2
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (T)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty