9-Fluoreneacetic acid ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2-(9H-Fluoren-9-yl)acetic acid
F0330-1G
TCF0330-001GEA
133.98
USD
TCF0330-001G
9-Fluoreneacetic acid ≥98.0% (by titrimetric analysis)
9-Fluoreneacetic acid
CAS Number: 6284-80-6
MDL Number: MFCD00013262
Molecular Formula: C15H12O2
Molecular Weight: 224.26
Purity/Analysis Method:
Form: Crystal
Formula:
C₁₅H₁₂O₂ MW: 224.26 g/mol |
MDL Number:
MFCD00013262 CAS Number: 6284-80-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...