Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Fluoroquinaldine ≥98.0%

Supplier: TCI America


F0300-5G
TCF0300-5GEA 100.92 USD InStock
TCF0300-5G
6-Fluoroquinaldine ≥98.0%
6-Fluoroquinaldine
CAS Number: 1128-61-6
MDL Number: MFCD00041233
Molecular Formula: C10H8FN
Molecular Weight: 161.18
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 99
Melting point (°C): 53
Flash Point (°C): 110
Storage Temperature: 0-10°C

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 6-Fluoroquinaldine


Formula: C₁₀H₈FN
MW: 161,18 g/mol
Boiling Pt: 99 °C
Melting Pt: 53 °C
Flash Pt: 110 °C
MDL Number: MFCD00041233
CAS Number: 1128-61-6
EINECS: 214-439-2
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty