1-Fluoro-3-iodobenzene ≥99.0% stabilized

Supplier: TCI America

Synonyms: 3-Fluoroiodobenzene

F0260-25G
TCF0260-25GEA 118 USD
TCF0260-25G
1-Fluoro-3-iodobenzene ≥99.0% stabilized
1-Fluoro-3-iodobenzene

(stabilized with Copper chip)
CAS Number: 1121-86-4
MDL Number: MFCD00001044
Molecular Formula: C6H4FI
Molecular Weight: 222.00
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 77
Flash Point (°C): 67
Specific Gravity (20/20): 1.92


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Benzyne Precursors (Aryne Precursors)
Other Product Names / Synonyms: 3-Fluoroiodobenzene

Formula: C₆H₄FI
MW: 222 g/mol
Boiling Pt: 77 °C
Density: 1.92 (20 °C)
Flash Pt: 67 °C
MDL Number: MFCD00001044
CAS Number: 1121-86-4
https://us.vwr.com/stibo/web/std.lang.all/88/59/28968859.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR