TCIAF0199

F0199-5G
TCF0199-5GEA 38.51 USD
TCF0199-5G
TCIAF0199
CAS Number: 6326-44-9
MDL Number: MFCD00003279
Molecular Formula: C8H13NO5
Molecular Weight: 203.19
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 174
Melting point (°C): 52
Flash Point (°C): 113

Other Product Names / Synonyms: Formaminomalonic Acid Diethyl Ester
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR