Formic acid rubidium salt ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Rubidium Formate

F0172-5G
TCF0172-005GEA 360 USD
TCF0172-005G
Formic acid rubidium salt ≥99.0% (by titrimetric analysis)
Formic acid rubidium salt

CAS Number: 3495-35-0
MDL Number: MFCD00054359
Molecular Formula: CH2O2
Molecular Weight: 130.48
Purity/Analysis Method: 99.0% (T)
Form: Crystal


Other Product Names / Synonyms: Rubidium Formate

Formula: CH₂O₂
MW: 130.48 g/mol
MDL Number: MFCD00054359
CAS Number: 3495-35-0
https://us.vwr.com/stibo/web/std.lang.all/87/85/28968785.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR