(Ferrocenylmethyl)trimethylammonium iodide ≥96.0% (by titrimetric analysis)
Synonyms:
(Ferrocenylmethyl)trimethylammonium Iodide
F0167-5G
TCF0167-005GEA
132.09
USD
TCF0167-005G
(Ferrocenylmethyl)trimethylammonium iodide ≥96.0% (by titrimetric analysis)
(Ferrocenylmethyl)trimethylammonium iodide
CAS Number: 12086-40-7
MDL Number: MFCD00041534
Molecular Formula: C14H20FeIN
Molecular Weight: 385.07
Purity/Analysis Method:
Form: Crystal
Formula:
C₁₄H₂₀FeIN MW: 385.07 g/mol |
MDL Number:
MFCD00041534 CAS Number: 12086-40-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...