Acid red 52
Supplier: TCI America
Synonyms:
Food Red No.106
F0143-25G
TCF0143-025GEA
128.91
USD
TCF0143-025G
Acid red 52
Acid red 52
CAS Number: 3520-42-1
MDL Number: MFCD00010180
Molecular Formula: C27H30N2O7S2
Molecular Weight: 580.65
Form: Powder
Color: Purple
Formula:
C₂₇H₃₀N₂O₇S₂ MW: 580.65 g/mol |
MDL Number:
MFCD00010180 CAS Number: 3520-42-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...