4-Fluoro-DL-phenylalanine ≥99.0% (by HPLC, titration analysis)
Supplier: TCI America
F0106-1G
TCF0106-001GEA
40.19
USD
TCF0106-001G
4-Fluoro-DL-phenylalanine ≥99.0% (by HPLC, titration analysis)
4-Fluoro-DL-phenylalanine
CAS Number: 51-65-0
MDL Number: MFCD00002600
Molecular Formula: C9H10FNO2
Molecular Weight: 183.18
Purity/Analysis Method:
Form: Crystal
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: 2-Amino-3-(4-fluorophenyl)propionic Acid, H-DL-Phe(4-F)-OH
Formula:
C₉H₁₀FNO₂ MW: 183.18 |
MDL Number:
MFCD00002600 CAS Number: 51-65-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...